Gimeracil 95%
Lieferant: Apollo Scientific
Synonyme:
5-Chloro-4-hydroxypyridin-2(1H)-one, 5-Chloro-4-hydroxy-2(1H)-pyridinone, 5-Chloro-2,4-pyridinediol, 5-Chloro-2,4-dihydroxypyridine
OR940679-100G
APOSOR940679-100GEA
126
EUR
APOSOR940679-100G
Gimeracil 95%
Gimeracil
Formel:
C₅H₄ClNO₂ Molecular Weight: 145,54 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08458352 CAS-Nummer: 103766-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...