2-Hydroxy-3-nitrobenzonitril
Lieferant: Apollo Scientific
|
Gefahr
|
OR940693-1G
OR940693-5G
APOSOR940693-1GEA
63.8
EUR
APOSOR940693-1G
APOSOR940693-5G
2-Hydroxy-3-nitrobenzonitril
2-Hydroxy-3-nitrobenzonitril
Formel:
C₇H₄N₂O₃ Molecular Weight: 164,1200 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234257 CAS-Nummer: 28177-79-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...