6-Methyl-1,3-benzoxazole-2(3H)-thione 95%

Lieferant: Apollo Scientific

Synonyme: 6-Methyl-3H-benzooxazole-2-thione

OR940696-1G OR940696-5G
APOSOR940696-1GEA 59.6 EUR
APOSOR940696-1G APOSOR940696-5G
6-Methyl-1,3-benzoxazole-2(3H)-thione 95%
6-Methyl-1,3-benzoxazole-2(3H)-thione
Formel: C₈H₇NOS
Molecular Weight: 165,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00175644
CAS-Nummer: 23417-29-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR