Methyl-4-brom-3-hydroxybenzoat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-bromo-3-hydroxybenzoate

OR9408-1G OR9408-25G OR9408-5G
APOSOR9408-1GEA 21.9 EUR
APOSOR9408-1G APOSOR9408-25G APOSOR9408-5G
Methyl-4-brom-3-hydroxybenzoat 97%
Methyl-4-brom-3-hydroxybenzoat
Formel: C₈H₇BrO₃
Molecular Weight: 231,0400 g/mol
Schmelzpunkt: 121…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08056372
CAS-Nummer: 106291-80-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR