Methyl-4-methyl-4-nitropentanoate 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 4-methyl-4-nitropentanoate
OR940897-1G
OR940897-5G
APOSOR940897-1GEA
92.8
EUR
APOSOR940897-1G
APOSOR940897-5G
Methyl-4-methyl-4-nitropentanoate 95%
Methyl-4-methyl-4-nitropentanoate
Formel:
C₇H₁₃NO₄ Molecular Weight: 175.19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047757 CAS-Nummer: 16507-02-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...