2,4-Dichlor-α-methylbenzylalkohol 95%

Lieferant: Apollo Scientific

Synonyme: 1-(2,4-Dichlorphenyl)ethanol, 2,4-Dichlorphenyl-methylcarbinol

OR941026-25G
APOSOR941026-25GEA 145 EUR
APOSOR941026-25G
2,4-Dichlor-α-methylbenzylalkohol 95%
2,4-Dichlor-α-methylbenzylalkohol
Formel: C₈H₈Cl₂O
Molecular Weight: 191,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004511
CAS-Nummer: 1475-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR