exo-2-Aminonorbornane 95%
Lieferant: Apollo Scientific
Synonyme:
(1R,2R,4S)-Bicyclo[2.2.1]heptan-2-amine, (1R,2R,4S)-rel-Bicyclo[2.2.1]heptan-2-amine, (1alpha,4alpha)-Norbornane-2alpha-amine
OR941069-1G
APOSOR941069-1GEA
75.4
EUR
APOSOR941069-1G
exo-2-Aminonorbornane 95%
exo-2-Aminonorbornane
Formel:
C₇H₁₃N Molecular Weight: 111,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078132 CAS-Nummer: 7242-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...