N-Benzyl-5-brom-2-methoxybenzolsulfonamid 97%

Lieferant: Apollo Scientific

OR9413-1G OR9413-5G
APOSOR9413-1GEA 139 EUR
APOSOR9413-1G APOSOR9413-5G
N-Benzyl-5-brom-2-methoxybenzolsulfonamid 97%
N-Benzyl-5-brom-2-methoxybenzolsulfonamid
Formel: C₁₄H₁₄BrNO₃S
Schmelzpunkt: 136…140 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02227932
CAS-Nummer: 446308-82-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR