4-Brom-3-hydroxybenzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR9414-5G
OR9414-100G
OR9414-25G
APOSOR9414-5GEA
87
EUR
APOSOR9414-5G
APOSOR9414-100G
APOSOR9414-25G
4-Brom-3-hydroxybenzoesäure 98%
4-Bromo-3-hydroxybenzoic acid
Formel:
C₇H₅BrO₃ Schmelzpunkt: 225…227 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08056371 CAS-Nummer: 14348-38-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...