Methyl-2-(m-tolyl)acetate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-methylphenylacetate

OR941614-5G
APOSOR941614-5GEA 98.6 EUR
APOSOR941614-5G
Methyl-2-(m-tolyl)acetate 95%
Methyl-2-(m-tolyl)acetate
Formel: C₁₀H₁₂O₂
Molecular Weight: 164,204 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00968468
CAS-Nummer: 53088-69-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR