2-Biphenylylisocyanat 95%
Lieferant: Apollo Scientific
Synonyme:
2-Biphenyl Isocyanate, Isocyanic acid 2-Biphenyl ester, 2-Biphenylylisocyanate
OR941696-5G
OR941696-25G
APOSOR941696-5GEA
121
EUR
APOSOR941696-5G
APOSOR941696-25G
2-Biphenylylisocyanat 95%
2-Biphenylylisocyanat
- May form small amount of sediment.
Formel:
C₁₃H₉NO Molecular Weight: 195.2167 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037087 CAS-Nummer: 17337-13-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...