6-Hydroxy-7-methoxy-4(1H)-chinazolinon 95%

Lieferant: Apollo Scientific

Synonyme: 6-Hydroxy-7-methoxy-4(3H)-quinazolinone, 6-Hydroxy-7-methoxy-3H-quinazolin-4-one

OR941735-10G OR941735-1G OR941735-5G
APOSOR941735-10GEA 63.8 EUR
APOSOR941735-10G APOSOR941735-1G APOSOR941735-5G
6-Hydroxy-7-methoxy-4(1H)-chinazolinon 95%
6-Hydroxy-7-methoxyquinazolin-4(1H)-one
Formel: C₉H₈N₂O₃
Molecular Weight: 192,1714 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20547768
CAS-Nummer: 179688-52-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR