3-(1-Pyrrolidinylsulphonyl)benzeneboronic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-(Pyrrolidin-1-ylsulphonyl)phenyl)boronic acid

OR9418-1G OR9418-10G OR9418-5G
APOSOR9418-1GEA 114 EUR
APOSOR9418-1G APOSOR9418-10G APOSOR9418-5G
3-(1-Pyrrolidinylsulphonyl)benzeneboronic acid 98%
3-(1-Pyrrolidinylsulphonyl)benzeneboronic acid
Formel: C₁₀H₁₄BNO₄S
Molecular Weight: 255.10 g/mol
Schmelzpunkt: 127…129 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07783851
CAS-Nummer: 871329-61-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR