Diallylmethylamine 95%

Lieferant: Apollo Scientific

OR941828-1G OR941828-5G
APOSOR941828-1GEA 139 EUR
APOSOR941828-1G APOSOR941828-5G
Diallylmethylamine 95%
Diallylmethylamine
Formel: C₇H₁₃N
Molecular Weight: 111.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00039824
CAS-Nummer: 2424-01-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR