Ethyl-5-amino-2-chlorbenzoat 95%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 5-amino-2-chlorobenzoate
OR942129-1G
OR942129-5G
APOSOR942129-1GEA
53.8
EUR
APOSOR942129-1G
APOSOR942129-5G
Ethyl-5-amino-2-chlorbenzoat 95%
Ethyl-5-amino-2-chlorbenzoat
Formel:
C₉H₁₀ClNO₂ Molecular Weight: 199,6342 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00115822 CAS-Nummer: 64401-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...