1-Methyl-1H-benzimidazol-6-amine 95%

Lieferant: Apollo Scientific

Synonyme: 1-Methyl-1H-benzo[d]imidazol-6-amine

OR942168-1G OR942168-250MG
APOSOR942168-1GEA 190 EUR
APOSOR942168-1G APOSOR942168-250MG
1-Methyl-1H-benzimidazol-6-amine 95%
1-Methyl-1H-benzimidazol-6-amine
Formel: C₈H₉N₃
Molecular Weight: 147.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09764050
CAS-Nummer: 26530-93-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR