N- (Triphenylmethyl)-DL-Serin-Methylester 98%

Lieferant: Apollo Scientific

Synonyme: N-(Triphenylmethyl)-DL-serine Methyl Ester, N-Trityl-DL-serine Methyl Ester, Trt-DL-Ser-OMe, Methyl 3-hydroxy-2-(tritylamino)propanoate

OR943085-25G OR943085-5G
APOSOR943085-25GEA 468 EUR
APOSOR943085-25G APOSOR943085-5G
N- (Triphenylmethyl)-DL-Serin-Methylester 98%
N- (Triphenylmethyl)-DL-Serin-Methylester
Formel: C₂₃H₂₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00583950
CAS-Nummer: 13515-76-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR