3-Amino-4-bromo-5-methylpyrazole 98%

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-5-methyl-1H-pyrazol-3-amine

OR943162-1G OR943162-25G OR943162-5G
APOSOR943162-1GEA 30.6 EUR
APOSOR943162-1G APOSOR943162-25G APOSOR943162-5G
3-Amino-4-bromo-5-methylpyrazole 98%
3-Amino-4-bromo-5-methylpyrazole
Formel: C₄H₆BrN₃
Molecular Weight: 176,02 g/mol
Schmelzpunkt: 117…122 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03412169
CAS-Nummer: 1780-72-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR