5-Oxo-1-phenylpyrrolidine-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR9432-1G OR9432-250MG OR9432-5G
APOSOR9432-1GEA 114 EUR
APOSOR9432-1G APOSOR9432-250MG APOSOR9432-5G
5-Oxo-1-phenylpyrrolidine-3-carboxylic acid
5-Oxo-1-phenylpyrrolidine-3-carboxylic acid
Formel: C₁₁H₁₁NO₃
Molecular Weight: 205.22 g/mol
Schmelzpunkt: 192…194 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00207656
CAS-Nummer: 39629-86-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR