2-Bromcyclohexanon 95%

Lieferant: Apollo Scientific

OR943232-1G OR943232-5G
APOSOR943232-1GEA 190 EUR
APOSOR943232-1G APOSOR943232-5G
2-Bromcyclohexanon 95%
2-Bromocyclohexanone
Formel: C₆H₉BrO
Lagertemperatur: Raumtemperatur
CAS-Nummer: 822-85-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR