1,4,7-Trimethyl-1,4,7-triazacyclononan 95%
Lieferant: Apollo Scientific
Synonyme:
1,4,7-Trimethyl-1,4,7-triazonane
OR943247-10G
OR943247-1G
OR943247-25G
OR943247-100G
APOSOR943247-10GEA
0
EUR
APOSOR943247-10G
APOSOR943247-1G
APOSOR943247-25G
APOSOR943247-100G
1,4,7-Trimethyl-1,4,7-triazacyclononan 95%
1,4,7-Trimethyl-1,4,7-triazacyclononane
Formel:
C₉H₂₁N₃ Molecular Weight: 171,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012359 CAS-Nummer: 96556-05-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...