[1-(3-Thienyl)ethyl]amine 95%
Lieferant: Apollo Scientific
Synonyme:
1-(Thiophen-3-yl)ethanamine
OR943265-1G
APOSOR943265-1GEA
125
EUR
APOSOR943265-1G
[1-(3-Thienyl)ethyl]amine 95%
[1-(3-Thienyl)ethyl]amine
Formel:
C₆H₉NS Molecular Weight: 127,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08452688 CAS-Nummer: 118488-08-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...