4,4'-Oxybis(1,4-phenylene)diboronic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,4'-Oxydibenzeneboronic acid, (Oxybis(4,1-phenylene))diboronic acid

OR9433-250MG OR9433-1G
APOSOR9433-250MGEA 401 EUR
APOSOR9433-250MG APOSOR9433-1G
4,4'-Oxybis(1,4-phenylene)diboronic acid 97%
4,4'-Oxybis(1,4-phenylene)diboronic acid
Formel: C₁₂H₁₂B₂O₅
Molecular Weight: 257,84 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00433207
CAS-Nummer: 19014-29-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR