2-Isopropyl-1,3-thiazole-4-carboxylic acid 95%
Lieferant: Apollo Scientific
Synonyme:
2-Isopropylthiazole-4-carboxylic acid
OR943371-1G
OR943371-250MG
APOSOR943371-1GEA
66.7
EUR
APOSOR943371-1G
APOSOR943371-250MG
2-Isopropyl-1,3-thiazole-4-carboxylic acid 95%
2-Isopropyl-1,3-thiazole-4-carboxylic acid
Formel:
C₇H₉NO₂S Molecular Weight: 171.22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09055228 CAS-Nummer: 234445-61-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...