(2,3-Dihydrothieno[3,4-b][1,4]dioxin-2-yl)methanol 95%

Lieferant: Apollo Scientific

Synonyme: Hydroxymethyl EDOT, EDT-methanol

OR943413-500MG OR943413-5G
APOSOR943413-500MGEA 0 EUR
APOSOR943413-500MG APOSOR943413-5G
(2,3-Dihydrothieno[3,4-b][1,4]dioxin-2-yl)methanol 95%
(2,3-Dihydrothieno[3,4-b][1,4]dioxin-2-yl)methanol
Formel: C₇H₈O₃S
Molecular Weight: 172,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11045357
CAS-Nummer: 146796-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR