1-(2-Chloroethyl)-2-pyrrolidone 95%

Lieferant: Apollo Scientific

Synonyme: 1-(2-chloroethyl)pyrrolidin-2-one, N-(beta-chloroethyl)-pyrrolidinone, 2-Pyrrolidinone, 1-(2-chloroethyl)-

OR943554-1G OR943554-250MG
APOSOR943554-1GEA 85.7 EUR
APOSOR943554-1G APOSOR943554-250MG
1-(2-Chloroethyl)-2-pyrrolidone 95%
1-(2-Chloroethyl)-2-pyrrolidone
Formel: C₆H₁₀ClNO
Molecular Weight: 147.6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00169064
CAS-Nummer: 51333-90-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR