1-(2,3-Dihydro-1,4-benzodioxin-6-yl)ethanamine 95%

Lieferant: Apollo Scientific

Synonyme: 1-(2,3-Dihydro-benzo[1,4]dioxin-6-yl)-ethylamine

OR943602-1G OR943602-250MG
APOSOR943602-1GEA 209 EUR
APOSOR943602-1G APOSOR943602-250MG
1-(2,3-Dihydro-1,4-benzodioxin-6-yl)ethanamine 95%
1-(2,3-Dihydro-1,4-benzodioxin-6-yl)ethanamine
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01312297
CAS-Nummer: 122416-41-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR