Monobenzyl malonate 95%

Lieferant: Apollo Scientific

Synonyme: 3-(benzyloxy)-3-oxopropanoic acid, 2-((Benzyloxy)carbonyl)acetic acid, malonic acid monobenzyl ester

OR944575-10G OR944575-1G OR944575-5G
APOSOR944575-10GEA 88.6 EUR
APOSOR944575-10G APOSOR944575-1G APOSOR944575-5G
Monobenzyl malonate 95%
Monobenzyl malonate
Formel: C₁₀H₁₀O₄
Molecular Weight: 194.186 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00138132
CAS-Nummer: 40204-26-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR