(4-Methoxy-3-nitrophenyl)methanol 95%

Lieferant: Apollo Scientific

OR944697-5G
APOSOR944697-5GEA 111 EUR
APOSOR944697-5G
(4-Methoxy-3-nitrophenyl)methanol 95%
(4-Methoxy-3-nitrophenyl)methanol
Formel: C₈H₉NO₄
Molecular Weight: 183,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02683526
CAS-Nummer: 41870-24-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR