4-(2-Pyrimidinyloxy)aniline 95%

Lieferant: Apollo Scientific

OR944926-1G OR944926-5G
APOSOR944926-1GEA 120 EUR
APOSOR944926-1G APOSOR944926-5G
4-(2-Pyrimidinyloxy)aniline 95%
4-(2-Pyrimidinyloxy)aniline
Formel: C₁₀H₉N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD01443685
CAS-Nummer: 105130-26-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR