Cyclopentane-1,1-dicarboxylic acid 95%

Lieferant: Apollo Scientific

OR945086-5G
APOSOR945086-5GEA 102 EUR
APOSOR945086-5G
Cyclopentane-1,1-dicarboxylic acid 95%
Cyclopentane-1,1-dicarboxylic acid
Formel: C₇H₁₀O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00029146
CAS-Nummer: 5802-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR