3,4-Dihydro-2H-1,4-benzoxazine-2-carboxylic acid 95%

Lieferant: Apollo Scientific

OR945249-1G OR945249-250MG
APOSOR945249-1GEA 440 EUR
APOSOR945249-1G APOSOR945249-250MG
3,4-Dihydro-2H-1,4-benzoxazine-2-carboxylic acid 95%
3,4-Dihydro-2H-1,4-benzoxazine-2-carboxylic acid
Formel: C₉H₉NO₃
Molecular Weight: 179.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05865108
CAS-Nummer: 90563-93-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR