1-(2-Naphthyl)ethanamine 95%
Lieferant: Apollo Scientific
Synonyme:
1-(Naphthalen-2-yl)ethanamine
OR945511-1G
APOSOR945511-1GEA
60.9
EUR
APOSOR945511-1G
1-(2-Naphthyl)ethanamine 95%
1-(2-Naphthyl)ethanamine
Formel:
C₁₂H₁₃N Molecular Weight: 171,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014321 CAS-Nummer: 1201-74-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...