4-Isopropyl-5-pyrimidinboronsäure 96%
Lieferant: Apollo Scientific
|
Achtung
|
OR9462-25MG
APOSOR9462-25MGEA
31.9
EUR
APOSOR9462-25MG
4-Isopropyl-5-pyrimidinboronsäure 96%
4-Isopropyl-5-pyrimidinboronsäure
Formel:
C₇H₁₁BN₂O₂ Molecular Weight: 165,99 g/mol Schmelzpunkt: 104…109 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08235084 CAS-Nummer: 913835-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...