4-(Chloromethyl)-N-methylbenzamide 98%
Lieferant: Apollo Scientific
OR946357-10G
OR946357-1G
OR946357-5G
APOSOR946357-10GEA
754
EUR
APOSOR946357-10G
APOSOR946357-1G
APOSOR946357-5G
4-(Chloromethyl)-N-methylbenzamide 98%
4-(Chloromethyl)-N-methylbenzamide
Formel:
C₉H₁₀ClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD11208365 CAS-Nummer: 220875-88-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...