3-Isopropoxybenzaldehyd 98%

Lieferant: Apollo Scientific

OR946447-1G OR946447-5G
APOSOR946447-1GEA 62.5 EUR
APOSOR946447-1G APOSOR946447-5G
3-Isopropoxybenzaldehyd 98%
3-Isopropoxybenzaldehyd
Formel: C₁₀H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01922005
CAS-Nummer: 75792-33-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR