5-Brom-2-methoxy-nicotinsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR9465-5G
OR9465-1G
OR9465-25G
APOSOR9465-5GEA
24.8
EUR
APOSOR9465-5G
APOSOR9465-1G
APOSOR9465-25G
5-Brom-2-methoxy-nicotinsäure 98%
5-Brom-2-methoxy-nicotinsäure
Formel:
C₇H₆BrNO₃ Molecular Weight: 232,04 g/mol Schmelzpunkt: 156…158 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06656771 CAS-Nummer: 54916-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...