Methyl-6-bromo-1-benzofuran-2-carboxylate 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 6-bromobenzofuran-2-carboxylate
OR946530-1G
OR946530-500MG
OR946530-5G
APOSOR946530-1GEA
336
EUR
APOSOR946530-1G
APOSOR946530-500MG
APOSOR946530-5G
Methyl-6-bromo-1-benzofuran-2-carboxylate 95%
Methyl-6-bromo-1-benzofuran-2-carboxylate
Formel:
C₁₀H₇BrO₃ Molecular Weight: 255,07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22126097 CAS-Nummer: 425675-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...