1-(tert-Butyl)-1H-pyrazol-4-amine 95%

Lieferant: Apollo Scientific

OR946666-1G OR946666-250MG
APOSOR946666-1GEA 98.6 EUR
APOSOR946666-1G APOSOR946666-250MG
1-(tert-Butyl)-1H-pyrazol-4-amine 95%
1-(tert-Butyl)-1H-pyrazol-4-amine
Formel: C₇H₁₃N₃
Molecular Weight: 139,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11845713
CAS-Nummer: 97421-13-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR