Ethyl-(2-aminophenyl)acetat 95%
Lieferant: Apollo Scientific
OR946796-1G
OR946796-250MG
APOSOR946796-1GEA
348
EUR
APOSOR946796-1G
APOSOR946796-250MG
Ethyl-(2-aminophenyl)acetat 95%
Ethyl-(2-aminophenyl)acetat
Formel:
C₁₀H₁₃NO₂ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
64460-85-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...