4-(3-Chlorpropylcarbamoyl)phenylboronsäure 97%

Lieferant: Apollo Scientific

Synonyme: (4-((3-Chloropropyl)carbamoyl)phenyl)boronic acid

OR9470-1G OR9470-5G
APOSOR9470-1GEA 125 EUR
APOSOR9470-1G APOSOR9470-5G
4-(3-Chlorpropylcarbamoyl)phenylboronsäure 97%
4-(3-Chlorpropylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₃BClNO₃
Molecular Weight: 241,4800 g/mol
Schmelzpunkt: 169…171 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235077
CAS-Nummer: 874460-03-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR