2-Hydroxy-5-methoxybenzonitril 95%
Lieferant: Apollo Scientific
OR947332-1G
OR947332-5G
APOSOR947332-1GEA
89.9
EUR
APOSOR947332-1G
APOSOR947332-5G
2-Hydroxy-5-methoxybenzonitril 95%
2-Hydroxy-5-methoxybenzonitril
Formel:
C₈H₇NO₂ Molecular Weight: 149,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02261935 CAS-Nummer: 39900-63-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...