(1-Isopropyl-3-piperidinyl)methanol 95%
Lieferant: Apollo Scientific
OR947764-1G
OR947764-5G
APOSOR947764-1GEA
319
EUR
APOSOR947764-1G
APOSOR947764-5G
(1-Isopropyl-3-piperidinyl)methanol 95%
(1-Isopropyl-3-piperidinyl)methanol
Formel:
C₉H₁₉NO Lagertemperatur: Raumtemperatur |
CAS-Nummer:
752970-45-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...