2-Cyano-3,6-dichloropyridine 95%

Lieferant: Apollo Scientific

Synonyme: 3,6-Dichloropicolinonitrile

OR947771-1G OR947771-250MG
APOSOR947771-1GEA 77 EUR
APOSOR947771-1G APOSOR947771-250MG
2-Cyano-3,6-dichloropyridine 95%
2-Cyano-3,6-dichloropyridine
Formel: C₆H₂Cl₂N₂
Molecular Weight: 173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00546824
CAS-Nummer: 1702-18-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR