FA-Gly-OH 95%

Lieferant: Apollo Scientific

OR947916-1G OR947916-5G
APOSOR947916-1GEA 190 EUR
APOSOR947916-1G APOSOR947916-5G
FA-Gly-OH 95%
FA-Gly-OH
Formel: C₉H₉NO₄
Molecular Weight: 195,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00080918
CAS-Nummer: 124882-74-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR