3-Oxo-3-(3-thienyl)propanenitrile 95%

Lieferant: Apollo Scientific

Synonyme: 3-Oxo-3-(thiophen-3-yl)propanenitrile

OR947992-1G OR947992-250MG
APOSOR947992-1GEA 87 EUR
APOSOR947992-1G APOSOR947992-250MG
3-Oxo-3-(3-thienyl)propanenitrile 95%
3-Oxo-3-(3-thienyl)propanenitrile
Formel: C₇H₅NOS
Lagertemperatur: Raumtemperatur
MDL: MFCD00111016
CAS-Nummer: 69879-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR