8-Oxo-5,6,7,8-tetrahydronaphthalene-2-carboxylic acid 95%

Lieferant: Apollo Scientific

OR948118-1G OR948118-250MG
APOSOR948118-1GEA 238 EUR
APOSOR948118-1G APOSOR948118-250MG
8-Oxo-5,6,7,8-tetrahydronaphthalene-2-carboxylic acid 95%
8-Oxo-5,6,7,8-tetrahydronaphthalene-2-carboxylic acid
Formel: C₁₁H₁₀O₃
Molecular Weight: 190.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00471460
CAS-Nummer: 89781-52-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR