(3-Chlorpropyl)(dimethoxy)methylsilan 95%
Lieferant: Apollo Scientific
Synonyme:
3-(Methyldimethoxysilyl)propyl Chloride, (3-Chloropropyl)dimethoxy(methyl)silane
OR948176-100G
APOSOR948176-100GEA
63.8
EUR
APOSOR948176-100G
(3-Chlorpropyl)(dimethoxy)methylsilan 95%
(3-Chlorpropyl)(dimethoxy)methylsilan
Formel:
C₆H₁₅ClO₂Si Molecular Weight: 182,72 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00053912 CAS-Nummer: 18171-19-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...