2-Chloro-5-iodo-4-methylpyridine 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR948230-5G
APOSOR948230-5GEA
121
EUR
APOSOR948230-5G
2-Chloro-5-iodo-4-methylpyridine 95%
2-Chloro-5-iodo-4-methylpyridine
Formel:
C₆H₅ClIN Lagertemperatur: Gefrierschrank |
MDL:
MFCD13185454 CAS-Nummer: 550347-54-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...