3-(2-Nitroethyl)phenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

OR9490-1G OR9490-250MG
APOSOR9490-1GEA 716 EUR
APOSOR9490-1G APOSOR9490-250MG
3-(2-Nitroethyl)phenylboronsäure 97%
3-(2-Nitroethyl)phenylboronsäure
Formel: C₈H₁₀BNO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD09258757
CAS-Nummer: 957034-42-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR